Assay Details
| Gene Symbol: RASSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637497.3 | XP_005637554.1 | 6 - 7 | 745 | 66 | |
| XM_844022.5 | XP_849115.2 | 6 - 7 | 732 | 66 | ||
More Information
Set Membership: |
|

RASSF4
Genomic Map