Assay Details
| Gene Symbol: DCTPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621286.3 | XP_005621343.1 | 284 | 76 | ||
| XM_547022.5 | XP_547022.1 | 1 | 288 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DCTPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621286.3 | XP_005621343.1 | 284 | 76 | ||
| XM_547022.5 | XP_547022.1 | 1 | 288 | 76 | ||
Set Membership: |
|