Assay Details
| Gene Symbol: PDE4DIPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630689.3 | XP_005630746.1 | 38 - 39 | 6380 | 117 | |
| XM_005630690.3 | XP_005630747.1 | 31 - 32 | 6632 | 117 | ||
| XM_005630691.3 | XP_005630748.1 | 37 - 38 | 6197 | 117 | ||
| XM_005630692.3 | XP_005630749.1 | 36 - 37 | 5807 | 117 | ||
| XM_014120591.2 | XP_013976066.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120592.2 | XP_013976067.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120593.2 | XP_013976068.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120594.2 | XP_013976069.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120595.2 | XP_013976070.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120596.2 | XP_013976071.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120597.2 | XP_013976072.1 | 38 - 39 | 6383 | 117 | ||
| XM_014120598.2 | XP_013976073.1 | 36 - 37 | 6044 | 117 | ||
| XM_014120599.2 | XP_013976074.1 | 38 - 39 | 6330 | 117 | ||
| XM_014120601.2 | XP_013976076.1 | 35 - 36 | 6002 | 117 | ||
| XM_014120602.2 | XP_013976077.1 | 37 - 38 | 6197 | 117 | ||
| XM_022404897.1 | XP_022260605.1 | 38 - 39 | 6383 | 117 | ||
| XM_022404898.1 | XP_022260606.1 | 31 - 32 | 6632 | 117 | ||
| XM_022404899.1 | XP_022260607.1 | 37 - 38 | 6197 | 117 | ||
More Information
Set Membership: |
|

PDE4DIPP2
Genomic Map