Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122645.1 | NP_001116117.1 | 1 - 2 | 371 | 85 | |
| GenBank mRNA | DN426900.1 | - | 1 - 2 | 148 | 85 | |
More Information
Set Membership: |
|

DIO2
Genomic Map