Assay Details
| Gene Symbol: SYNJ2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252343.1 | NP_001239272.1 | 2 - 3 | 237 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYNJ2BP| Gene Symbol: SYNJ2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252343.1 | NP_001239272.1 | 2 - 3 | 237 | 102 | |
Set Membership: |
|