Assay Details
| Gene Symbol: ATP5J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621086.3 | XP_005621143.1 | 912 | 85 | ||
| XM_014114252.1 | XP_013969727.1 | 843 | 85 | |||
| XM_014114254.2 | XP_013969729.2 | 702 | 85 | |||
| XM_014114255.2 | XP_013969730.2 | 661 | 85 | |||
| XM_022419690.1 | XP_022275398.1 | 843 | 85 | |||
| XM_022419691.1 | XP_022275399.1 | 765 | 85 | |||
| XM_022419692.1 | XP_022275400.1 | 754 | 85 | |||
| XM_022419693.1 | XP_022275401.1 | 708 | 85 | |||
| XM_022419694.1 | XP_022275402.1 | 739 | 85 | |||
More Information
Set Membership: |
|

ATP5J2
Genomic Map