Assay Details
| Gene Symbol: LOC489707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620685.3 | XP_005620742.1 | 2 - 3 | 306 | 86 | |
| XM_005620686.1 | XP_005620743.1 | 2 - 3 | 397 | 86 | ||
| XM_005620687.3 | XP_005620744.1 | 2 - 3 | 236 | 86 | ||
| XM_005620688.2 | XP_005620745.1 | 2 - 3 | 412 | 86 | ||
| XM_005620690.3 | XP_005620747.1 | 3 - 4 | 381 | 86 | ||
| XM_022419179.1 | XP_022274887.1 | 2 - 3 | 154 | 86 | ||
| XM_022419180.1 | XP_022274888.1 | 2 - 3 | 164 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005620685.3, XM_005620686.1, XM_005620687.3, XM_005620688.2, XM_005620690.3, XM_022419179.1, XM_022419180.1 transcripts.
Set Membership: |
|

LOC489707
Genomic Map