Assay Details
| Gene Symbol: LOC606786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113267.2 | XP_013968742.1 | 782 | 54 | ||
| XM_022418064.1 | XP_022273772.1 | 782 | 54 | |||
| XM_022418065.1 | XP_022273773.1 | 1048 | 54 | |||
| XM_022418066.1 | XP_022273774.1 | 672 | 54 | |||
| XM_843133.4 | XP_848226.1 | 781 | 54 | |||
More Information
Set Membership: |
|

LOC606786
Genomic Map