Assay Details
| Gene Symbol: RP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111448.2 | XP_013966923.1 | 2 - 3 | 955 | 143 | |
| XM_548972.6 | XP_548972.2 | 2 - 3 | 956 | 143 | ||
More Information
Set Membership: |
|

RP2
Genomic Map