Assay Details
| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195695.1 | NP_001182624.1 | 663 | 102 | ||
| XM_005621225.3 | XP_005621282.1 | 757 | 102 | |||
| XM_022419489.1 | XP_022275197.1 | 757 | 102 | |||
| XM_022419490.1 | XP_022275198.1 | 484 | 102 | |||
| GenBank mRNA | DN748234.1 | - | 196 | 102 | ||
More Information
Set Membership: |
|

CCDC189
Genomic Map