Assay Details
| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195695.1 | NP_001182624.1 | 930 | 74 | ||
| GenBank mRNA | DN748234.1 | - | 463 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC189| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195695.1 | NP_001182624.1 | 930 | 74 | ||
| GenBank mRNA | DN748234.1 | - | 463 | 74 | ||
Set Membership: |
|