Assay Details
| Gene Symbol: PRR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419851.1 | XP_022275559.1 | 1845 | 111 | ||
| XM_022419852.1 | XP_022275560.1 | 1764 | 111 | |||
| XM_844097.5 | XP_849190.1 | 1838 | 111 | |||
More Information
Set Membership: |
|

PRR14
Genomic Map