Assay Details
| Gene Symbol: LOC489748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620802.3 | XP_005620859.1 | 7 - 8 | 1072 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC489748| Gene Symbol: LOC489748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620802.3 | XP_005620859.1 | 7 - 8 | 1072 | 98 | |
Set Membership: |
|