Assay Details
| Gene Symbol: LOC489812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114176.2 | XP_013969651.2 | 3387 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC489812| Gene Symbol: LOC489812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114176.2 | XP_013969651.2 | 3387 | 71 | ||
Set Membership: |
|