Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624376.3 | XP_005624433.1 | 807 | 73 | ||
| XM_022422887.1 | XP_022278595.1 | 808 | 73 | |||
| XM_843285.5 | XP_848378.1 | 806 | 73 | |||
| GenBank mRNA | KR827462.1 | - | 784 | 73 | ||
| KR827463.1 | - | 784 | 73 | |||
| KR827464.1 | - | 784 | 73 | |||
| KT192567.1 | - | 548 | 73 | |||
More Information
Set Membership: |
|

GFAP
Genomic Map