Assay Details
| Gene Symbol: TP53BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622650.3 | XP_005622707.1 | 3 - 4 | 337 | 70 | |
| XM_005622652.3 | XP_005622709.1 | 4 - 5 | 434 | 70 | ||
| XM_014115274.2 | XP_013970749.1 | 3 - 4 | 337 | 70 | ||
| XM_014115275.2 | XP_013970750.1 | 5 - 6 | 497 | 70 | ||
More Information
Set Membership: |
|

TP53BP2
Genomic Map