Assay Details
| Gene Symbol: SETD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623792.3 | XP_005623849.1 | 2 - 3 | 260 | 58 | |
| XM_022422370.1 | XP_022278078.1 | 2 - 3 | 134 | 58 | ||
| XM_547974.6 | XP_547974.2 | 2 - 3 | 258 | 58 | ||
More Information
Set Membership: |
|

SETD3
Genomic Map