Assay Details
| Gene Symbol: RIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115804.2 | XP_013971279.1 | 3 - 4 | 917 | 77 | |
| XM_014115805.2 | XP_013971280.1 | 3 - 4 | 486 | 77 | ||
| XM_014115807.2 | XP_013971282.1 | 4 - 5 | 644 | 77 | ||
| XM_022421858.1 | XP_022277566.1 | 4 - 5 | 792 | 77 | ||
| XM_022421860.1 | XP_022277568.1 | 4 - 5 | 794 | 77 | ||
| XM_022421861.1 | XP_022277569.1 | 4 - 5 | 797 | 77 | ||
More Information
Set Membership: |
|

RIPK3
Genomic Map