Assay Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427149.1 | XP_022282857.1 | 4 - 5 | 691 | 73 | |
| XM_022427152.1 | XP_022282860.1 | 4 - 5 | 680 | 73 | ||
| XM_845443.5 | XP_850536.1 | 4 - 5 | 691 | 73 | ||
More Information
Set Membership: |
|

CHMP2A
Genomic Map