Assay Details
| Gene Symbol: ECHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615626.3 | XP_005615683.1 | 2 - 3 | 478 | 99 | |
| XM_005615627.3 | XP_005615684.1 | 2 - 3 | 368 | 99 | ||
| XM_005615628.2 | XP_005615685.1 | 2 - 3 | 280 | 99 | ||
| XM_022419841.1 | XP_022275549.1 | 2 - 3 | 482 | 99 | ||
More Information
Set Membership: |
|

ECHDC1
Genomic Map