Assay Details
| Gene Symbol: EPB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412615.1 | XP_022268323.1 | 8 - 9 | 1659 | 82 | |
| XM_022412616.1 | XP_022268324.1 | 9 - 10 | 1765 | 82 | ||
| XM_022412617.1 | XP_022268325.1 | 9 - 10 | 1766 | 82 | ||
| XM_846088.6 | XP_851181.3 | 9 - 10 | 1763 | 82 | ||
More Information
Set Membership: |
|

EPB42
Genomic Map