Assay Details
| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_541197.5 | XP_541197.2 | 2 - 3 | 265 | 61 | |
| XM_846017.4 | XP_851110.1 | 2 - 3 | 345 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_541197.5, XM_846017.4 transcripts.
Set Membership: |
|

CCR6
Genomic Map