Assay Details
| Gene Symbol: PARP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639565.3 | XP_005639622.1 | 8 - 9 | 1641 | 123 | |
| XM_005639568.3 | XP_005639625.1 | 5 - 6 | 1073 | 123 | ||
| XM_014110294.2 | XP_013965769.1 | 7 - 8 | 1531 | 123 | ||
| XM_022413832.1 | XP_022269540.1 | 5 - 6 | 673 | 123 | ||
| XM_022413833.1 | XP_022269541.1 | 7 - 8 | 1567 | 123 | ||
More Information
Set Membership: |
|

PARP15
Genomic Map