Assay Details
| Gene Symbol: PARP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639565.3 | XP_005639622.1 | 9 - 10 | 1789 | 85 | |
| XM_005639568.3 | XP_005639625.1 | 6 - 7 | 1221 | 85 | ||
| XM_014110294.2 | XP_013965769.1 | 8 - 9 | 1679 | 85 | ||
| XM_022413832.1 | XP_022269540.1 | 6 - 7 | 821 | 85 | ||
| XM_022413833.1 | XP_022269541.1 | 8 - 9 | 1715 | 85 | ||
| XM_022413834.1 | XP_022269542.1 | 7 - 8 | 1458 | 85 | ||
More Information
Set Membership: |
|

PARP15
Genomic Map