Assay Details
| Gene Symbol: BABAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630287.2 | XP_005630344.1 | 7 - 8 | 852 | 86 | |
| XM_022404584.1 | XP_022260292.1 | 7 - 8 | 836 | 86 | ||
| XM_022404585.1 | XP_022260293.1 | 7 - 8 | 852 | 86 | ||
| XM_022404586.1 | XP_022260294.1 | 7 - 8 | 852 | 86 | ||
| XM_022404587.1 | XP_022260295.1 | 7 - 8 | 852 | 86 | ||
| XM_022404588.1 | XP_022260296.1 | 7 - 8 | 852 | 86 | ||
| XM_845588.4 | XP_850681.1 | 7 - 8 | 852 | 86 | ||
More Information
Set Membership: |
|

BABAM2
Genomic Map