Assay Details
| Gene Symbol: CR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622317.2 | XP_005622374.1 | 2629 | 56 | ||
| XM_022420856.1 | XP_022276564.1 | 2629 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622317.2 | XP_005622374.1 | 2629 | 56 | ||
| XM_022420856.1 | XP_022276564.1 | 2629 | 56 | |||
Set Membership: |
|