Assay Details
| Gene Symbol: FOLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633554.3 | XP_005633611.1 | 3 - 4 | 447 | 57 | |
| XM_022407744.1 | XP_022263452.1 | 3 - 4 | 478 | 57 | ||
| XM_846900.5 | XP_851993.1 | 2 - 3 | 353 | 57 | ||
More Information
Set Membership: |
|

FOLR1
Genomic Map