Assay Details
| Gene Symbol: BLOC1S5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110576.2 | XP_013966051.1 | 3 - 4 | 297 | 96 | |
| XM_846732.6 | XP_851825.3 | 4 - 5 | 426 | 96 | ||
More Information
Set Membership: |
|

BLOC1S5
Genomic Map