Assay Details
| Gene Symbol: ANGPT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634894.3 | XP_005634951.1 | 8 - 9 | 2032 | 91 | |
| XM_014107142.2 | XP_013962617.1 | 8 - 9 | 2032 | 91 | ||
| XM_022408848.1 | XP_022264556.1 | 8 - 9 | 2032 | 91 | ||
| XM_846919.5 | XP_852012.1 | 8 - 9 | 2031 | 91 | ||
More Information
Set Membership: |
|

ANGPT4
Genomic Map