Assay Details
| Gene Symbol: CARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631355.2 | XP_005631412.1 | 5 - 6 | 488 | 62 | |
| XM_846131.4 | XP_851224.1 | 6 - 7 | 742 | 62 | ||
More Information
Set Membership: |
|

CARS1
Genomic Map