Assay Details
| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639547.2 | XP_005639604.2 | 4 - 5 | 492 | 76 | |
| XM_022413917.1 | XP_022269625.1 | 4 - 5 | 492 | 76 | ||
| XM_022413918.1 | XP_022269626.1 | 4 - 5 | 492 | 76 | ||
| XM_022413920.1 | XP_022269628.1 | 4 - 5 | 492 | 76 | ||
| XM_845479.4 | XP_850572.3 | 4 - 5 | 492 | 76 | ||
More Information
Set Membership: |
|

RABL3
Genomic Map