Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114757.1 | NP_001108229.1 | 3 - 4 | 316 | 87 | |
| GenBank mRNA | DN370248.1 | - | 3 - 4 | 313 | 87 | |
More Information
Set Membership: |
|

SELENOS
Genomic Map