Assay Details
| Gene Symbol: GRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625856.3 | XP_005625913.2 | 6 - 7 | 1017 | 70 | |
| XM_005625859.3 | XP_005625916.1 | 6 - 7 | 881 | 70 | ||
| XM_022424399.1 | XP_022280107.1 | 6 - 7 | 768 | 70 | ||
| XM_022424400.1 | XP_022280108.1 | 5 - 6 | 1490 | 70 | ||
| XM_022424401.1 | XP_022280109.1 | 4 - 5 | 601 | 70 | ||
| XM_844613.4 | XP_849706.1 | 6 - 7 | 940 | 70 | ||
More Information
Set Membership: |
|

GRAP2
Genomic Map