Assay Details
| Gene Symbol: DTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107079.2 | XP_013962554.1 | 3 - 4 | 487 | 61 | |
| XM_014107080.2 | XP_013962555.1 | 3 - 4 | 487 | 61 | ||
| XM_022409425.1 | XP_022265133.1 | 3 - 4 | 487 | 61 | ||
| XM_022409426.1 | XP_022265134.1 | 3 - 4 | 487 | 61 | ||
More Information
Set Membership: |
|

DTD1
Genomic Map