Assay Details
| Gene Symbol: TANGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410292.1 | XP_022266000.1 | 6 - 7 | 514 | 63 | |
| XM_022410293.1 | XP_022266001.1 | 5 - 6 | 489 | 63 | ||
| XM_022410294.1 | XP_022266002.1 | 6 - 7 | 696 | 63 | ||
More Information
Set Membership: |
|

TANGO2
Genomic Map