Assay Details
| Gene Symbol: ANXA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630779.3 | XP_005630836.1 | 5 - 6 | 553 | 60 | |
| XM_005630781.3 | XP_005630838.1 | 5 - 6 | 554 | 60 | ||
| XM_014120650.2 | XP_013976125.1 | 5 - 6 | 553 | 60 | ||
| XM_022404980.1 | XP_022260688.1 | 5 - 6 | 550 | 60 | ||
| XM_022404981.1 | XP_022260689.1 | 5 - 6 | 546 | 60 | ||
| XM_022404982.1 | XP_022260690.1 | 5 - 6 | 578 | 60 | ||
| XM_022404983.1 | XP_022260691.1 | 5 - 6 | 553 | 60 | ||
| XM_022404984.1 | XP_022260692.1 | 4 - 5 | 464 | 60 | ||
| XM_022404985.1 | XP_022260693.1 | 4 - 5 | 556 | 60 | ||
More Information
Set Membership: |
|

ANXA9
Genomic Map