Assay Details
| Gene Symbol: MSRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107567.2 | XP_013963042.1 | 5 - 6 | 930 | 67 | |
| XM_014107568.1 | XP_013963043.1 | 5 - 6 | 422 | 67 | ||
| XM_014107569.2 | XP_013963044.1 | 5 - 6 | 434 | 67 | ||
| XM_014107570.2 | XP_013963045.1 | 5 - 6 | 835 | 67 | ||
| XM_014107571.2 | XP_013963046.1 | 5 - 6 | 570 | 67 | ||
| XM_014107572.2 | XP_013963047.1 | 6 - 7 | 903 | 67 | ||
| XM_022409753.1 | XP_022265461.1 | 4 - 5 | 507 | 67 | ||
More Information
Set Membership: |
|

MSRA
Genomic Map