Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433604.4 | XP_003433652.1 | 2 - 3 | 441 | 91 | |
| XM_845909.4 | XP_851002.1 | 2 - 3 | 656 | 91 | ||
More Information
Set Membership: |
|

AVPI1
Genomic Map