Assay Details
| Gene Symbol: SAP30BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116280.2 | XP_013971755.1 | 342 | 67 | ||
| XM_022422723.1 | XP_022278431.1 | 354 | 67 | |||
| XM_844378.5 | XP_849471.1 | 387 | 67 | |||
More Information
Set Membership: |
|

SAP30BP
Genomic Map