Assay Details
| Gene Symbol: AAAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636772.3 | XP_005636829.1 | 5 - 6 | 379 | 58 | |
| XM_005636773.3 | XP_005636830.1 | 5 - 6 | 420 | 58 | ||
| XM_014108424.2 | XP_013963899.1 | 5 - 6 | 471 | 58 | ||
| XM_844704.6 | XP_849797.3 | 5 - 6 | 560 | 58 | ||
More Information
Set Membership: |
|

AAAS
Genomic Map