Assay Details
| Gene Symbol: IAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639605.4 | XP_003639653.1 | 358 | 76 | ||
| XM_022404433.1 | XP_022260141.1 | 359 | 76 | |||
| XM_022404434.1 | XP_022260142.1 | 359 | 76 | |||
| XM_022404435.1 | XP_022260143.1 | 232 | 76 | |||
| XM_022404436.1 | XP_022260144.1 | 264 | 76 | |||
| XM_022404437.1 | XP_022260145.1 | 261 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022404435.1, XM_022404436.1, XM_022404437.1 transcripts.
Set Membership: |
|

IAH1
Genomic Map