Assay Details
| Gene Symbol: SYNGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252581.1 | NP_001239510.1 | 497 | 70 | ||
| GenBank mRNA | DN269979.1 | - | 304 | 70 | ||
| DN418042.1 | - | 497 | 70 | |||
More Information
Set Membership: |
|

SYNGR2
Genomic Map