Assay Details
| Gene Symbol: YAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120832.2 | XP_013976307.1 | 179 | 78 | ||
| XM_022405180.1 | XP_022260888.1 | 179 | 78 | |||
| XM_846301.3 | XP_851394.1 | 179 | 78 | |||
More Information
Set Membership: |
|

YAE1
Genomic Map