Assay Details
| Gene Symbol: TBATA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417532.1 | XP_022273240.1 | 9 - 10 | 1239 | 77 | |
| XM_022417533.1 | XP_022273241.1 | 9 - 10 | 1239 | 77 | ||
| XM_022417534.1 | XP_022273242.1 | 7 - 8 | 993 | 77 | ||
| XM_022417535.1 | XP_022273243.1 | 8 - 9 | 1065 | 77 | ||
| XM_022417536.1 | XP_022273244.1 | 8 - 9 | 996 | 77 | ||
| XM_022417537.1 | XP_022273245.1 | 6 - 7 | 786 | 77 | ||
| XM_022417539.1 | XP_022273247.1 | 8 - 9 | 963 | 77 | ||
More Information
Set Membership: |
|

TBATA
Genomic Map