Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635975.3 | XP_005636032.2 | 7 - 8 | 762 | 125 | |
| XM_846816.6 | XP_851909.2 | 7 - 8 | 716 | 125 | ||
More Information
Set Membership: |
|

SEPTIN2
Genomic Map