Assay Details
| Gene Symbol: CHST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631305.3 | XP_005631362.1 | 2 - 3 | 1195 | 153 | |
| GenBank mRNA | FR775794.1 | - | 3 - 4 | 406 | 153 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005631305.3 transcripts.
Set Membership: |
|

CHST1
Genomic Map