Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405905.1 | XP_022261613.1 | 236 | 94 | ||
XM_844835.5 | XP_849928.1 | 353 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
