Assay Details
| Gene Symbol: UPK3BL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_844944.4 | XP_850037.1 | 714 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPK3BL2| Gene Symbol: UPK3BL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_844944.4 | XP_850037.1 | 714 | 82 | ||