Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631200.3 | XP_005631257.1 | 2 - 3 | 992 | 80 | |
| XM_005631201.3 | XP_005631258.1 | 2 - 3 | 944 | 80 | ||
| XM_847579.5 | XP_852672.1 | 2 - 3 | 795 | 80 | ||
More Information
Set Membership: |
|

CLP1
Genomic Map