Assay Details
| Gene Symbol: LOC484411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616363.3 | XP_005616420.1 | 3 - 4 | 231 | 100 | |
| XM_005616364.3 | XP_005616421.1 | 4 - 5 | 284 | 100 | ||
| XM_005616365.3 | XP_005616422.1 | 3 - 4 | 230 | 100 | ||
| XM_022424383.1 | XP_022280091.1 | 3 - 4 | 232 | 100 | ||
| Gene Symbol: CARD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424373.1 | XP_022280081.1 | 4 - 5 | 611 | 100 | |
| XM_022424375.1 | XP_022280083.1 | 5 - 6 | 673 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005616363.3, XM_005616364.3, XM_005616365.3, XM_022424373.1, XM_022424375.1, XM_022424383.1 transcripts.
Set Membership: |
|

LOC484411
Genomic Map